Copper(II) hexafluoroacetylacetonate hydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hexafluoroacetylacetono Copper(II) Salt, Copper(II) Hexafluoroacetylacetonate, Copper(II) hexafluoro-2,4-pentanedionate hydrate, Bis(hexafluoroacetylacetonato)copper(II) hydrate, Copper(II) hexafluoro-2,4-pentanedionate xhydrate

B25284-06 B25284-14
CAAAAB25284-06EA 111.23 CAD
CAAAAB25284-06 CAAAAB25284-14
Copper(II) hexafluoroacetylacetonate hydrate 98%
Copper (II) hexafluoroacetylacetonate hydrate

Purity: 98% CAS No.: 155640-85-0 Molecular Formula: C10H2CuF12O4.xH2O Formula Weight: 477.64(anhy) Melting Point: ca 100° Boiling Point: 220° MDL No.: MFCD00151019

Formula: C₁₀H₂CuF₁₂O₄·xH₂O
MW: 477.64 g/mol (anhy)
Boiling Pt: 220 °C
Melting Pt: ∼ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00151019
CAS Number: 155640-85-0
EINECS: 238-850-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR