1-Tritylimidazole-4-carbaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Trityl-1H-imidazole-4-carboxaldehyde, 1-Trityl-1H-imidazole-4-carbaldehyde, 1-Tritylimidazole-4-carboxaldehyde
B25272-06
B25272-14
CAAAAB25272-03EA
90.79
CAD
CAAAAB25272-03
CAAAAB25272-06
CAAAAB25272-14
1-Tritylimidazole-4-carbaldehyde 98%
1-Tritylimidazole-4-carbaldehyde
Purity: 98%
CAS No.: 33016-47-6
Molecular Formula: C23H18N2O
Formula Weight: 338.41
Melting Point: ca 192° dec.
MDL No.: MFCD02179554
Formula:
C₂₃H₁₈N₂O MW: 338.41 g/mol Melting Pt: ca 192 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD02179554 CAS Number: 33016-47-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...