2,4-Dichlorobenzylideneacetone 97%
Supplier: Thermo Scientific Chemicals
B25270-03
B25270-06
CAAAAB25270-03EA
35.87
CAD
CAAAAB25270-03
CAAAAB25270-06
CAAAAB25270-14
2,4-Dichlorobenzylideneacetone 97%
2,4-Dichlorobenzylideneacetone
Purity: 97%
CAS No.: 61888-78-6
Molecular Formula: C10H8Cl2O
Formula Weight: 215.08
Melting Point: 83-85°
MDL No.: MFCD00052852
Formula:
C₁₀H₈Cl₂O Melting Pt: 83…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052852 CAS Number: 61888-78-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...