Methyl-2-oxocyclopentanecarboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

B25256-06 B25256-14 B25256-22
CAAAAB25256-06EA 46.77 CAD
CAAAAB25256-06 CAAAAB25256-14 CAAAAB25256-22
Methyl-2-oxocyclopentanecarboxylate 97%
Methyl 2-oxocyclopentanecarboxylate

Purity: 97% CAS No.: 10472-24-9 Molecular Formula: C7H10O3 Formula Weight: 142.16 Flash Point: 110°(230°F) Refractive Index: 1.4565 Boiling Point: 105-107°/20mm MDL No.: MFCD00001411

Formula: C₇H₁₀O₃
Boiling Pt: 105…107 °C/20 mm
Density: 1.145
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001411
CAS Number: 10472-24-9
EINECS: 233-962-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR