4-(Chloromethyl)-1,2-diphenylethane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B25222-03
B25222-06
B25222-14
CAAAAB25222-03EA
92.27
CAD
CAAAAB25222-03
CAAAAB25222-06
CAAAAB25222-14
4-(Chloromethyl)-1,2-diphenylethane 98%
Purity: 98%
CAS No.: 80676-35-3
Molecular Formula: C15H15Cl
Formula Weight: 230.73
Melting Point: 38-42°
UN No.: 1759
MDL No.: MFCD02093918
Formula:
C₁₅H₁₅Cl Melting Pt: 38…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093918 CAS Number: 80676-35-3 UN: 1759 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...