2-Ethyl-6-methylphenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25213-03
B25213-06
B25213-14
CAAAAB25213-03EA
59.72
CAD
CAAAAB25213-03
CAAAAB25213-06
CAAAAB25213-14
2-Ethyl-6-methylphenol 97%
2-Ethyl-6-methylphenol
Purity: 97%
CAS No.: 1687-64-5
Molecular Formula: C9H12O
Formula Weight: 136.19
Refractive Index: 1.5310
Formula:
C₉H₁₂O Storage Temperature: Ambient |
MDL Number:
MFCD00962292 CAS Number: 1687-64-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...