Quinoline-N-oxide hydrate, (∼20% H₂O) 97% (dry weight)
Supplier: Thermo Scientific Chemicals
B25209-06
B25209-14
CAAAAB25209-06EA
98.12
CAD
CAAAAB25209-06
CAAAAB25209-14
Quinoline-N-oxide hydrate, (∼20% H₂O) 97% (dry weight)
Quinoline-N-oxide hydrate
Purity: 97%
CAS No.: 64201-64-5
Molecular Formula: C9H7NO.xH2O
Formula Weight: 145.16(anhy)
Flash Point:
Formula:
C₉H₇NO·xH₂O Melting Pt: ca 50 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00149472 CAS Number: 64201-64-5 EINECS: 216-560-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...