Calcium pyruvate, anhydrous 97%
Supplier: Thermo Scientific Chemicals
B25204-14
B25204-22
CAAAAB25204-14EA
68.89
CAD
CAAAAB25204-14
CAAAAB25204-22
CAAAAB25204-36
Calcium pyruvate, anhydrous 97%
Calcium pyruvate
Purity: 97%
CAS No.: 52009-14-0
Molecular Formula: C6H6CaO6
Formula Weight: 214.19
MDL No.: MFCD00037199
Formula:
C₆H₆CaO₆ Storage Temperature: Ambient |
MDL Number:
MFCD00037199 CAS Number: 52009-14-0 EINECS: 257-599-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...