5-tert-Butyl-2-iodo-1,3-dimethylbenzene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25195-03
B25195-06
CAAAAB25195-03EA
89.27
CAD
CAAAAB25195-03
CAAAAB25195-06
CAAAAB25195-14
5-tert-Butyl-2-iodo-1,3-dimethylbenzene 97%
5-tert-Butyl-2-iodo-1,3-dimethylbenzene
Purity: 97%
CAS No.: 5122-20-3
Molecular Formula: C12H17I
Formula Weight: 288.17
Odor: Odorless
Melting Point: 54-57°
MDL No.: MFCD00052173
Formula:
C₁₂H₁₇I Melting Pt: 54…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052173 CAS Number: 5122-20-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...