5-Bromo-2-fluorobenzylamine hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(5-Bromo-2-fluorophenyl)methanamine hydrochloride
B25176-06
B25176-14
CAAAAB25176-06EA
68.89
CAD
CAAAAB25176-06
CAAAAB25176-14
AAB25176-03
5-Bromo-2-fluorobenzylamine hydrochloride 97%
5-Bromo-2-fluorobenzylamine hydrochloride
Purity: 97%
CAS No.: 202865-69-8
Molecular Formula: C7H7BrFN.HCl
Formula Weight: 240.50
Melting Point: 221-226°
MDL No.: MFCD00143426
Formula:
C₇H₈BrClFN MW: 240.50 g/mol Melting Pt: 221…226 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143426 CAS Number: 202865-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...