Methyl isoxazole-5-carboxylate 97%

Supplier: Thermo Scientific Chemicals

B25169-03 B25169-06
CAAAB25169-03EA 183.28 CAD
CAAAB25169-03 CAAAB25169-06 CAAAB25169-14
Methyl isoxazole-5-carboxylate 97%
Methyl isoxazole-5-carboxylate

Purity: 97% CAS No.: 15055-81-9 Molecular Formula: C5H5NO3 Formula Weight: 127.10 Melting Point: 48-50° MDL No.: MFCD01570794

Formula: C₅H₅NO₃
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD01570794
CAS Number: 15055-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR