Propyl decanoate 98%

Supplier: Thermo Scientific Chemicals

B25159-0B B25159-18 B25159-30
CAAAAB25159-0BEA 597.24 CAD
CAAAAB25159-0B CAAAAB25159-18 CAAAAB25159-30
Propyl decanoate 98%
Propyl decanoate

Purity: 98% CAS No.: 30673-60-0 Molecular Formula: CH3(CH2)8CO2CH2CH2CH3 Formula Weight: 214.35 Flash Point: 115°(239°F) Boiling Point: 254° MDL No.: MFCD00056329

Formula: C₁₃H₂₆O₂
MW: 214.35 g/mol
Boiling Pt: 254 °C
Density: 0.86
Flash Pt: 115 °C (239 °F)
Storage Temperature: Ambient
MDL Number: MFCD00056329
CAS Number: 30673-60-0
EINECS: 250-283-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR