1-(2-Phenylethyl)piperazine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B25158-09
B25158-18
CAAAAB25158-09EA
81.84
CAD
CAAAAB25158-09
CAAAAB25158-18
CAAAAB25158-30
1-(2-Phenylethyl)piperazine 98%
1-(2-Phenylethyl)piperazine
Purity: 98%
CAS No.: 5321-49-3
Molecular Formula: C12H18N2
Formula Weight: 190.29
Flash Point:
Formula:
C₁₂H₁₈N₂ Boiling Pt: 276 °C Density: 1.01 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040740 CAS Number: 5321-49-3 EINECS: 226-186-5 UN: 2735 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...