4-(4-Hydroxy-3-methoxyphenyl)-2-butanone 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Zingerone, Vanillylacetone

B25150-03 B25150-06 B25150-14
CAAAAB25150-03EA 58.3 CAD
CAAAAB25150-03 CAAAAB25150-06 CAAAAB25150-14
4-(4-Hydroxy-3-methoxyphenyl)-2-butanone 97%
4-(4-Hydroxy-3-methoxyphenyl)-2-butanone

Purity: 97% CAS No.: 122-48-5 Molecular Formula: C11H14O3 Formula Weight: 194.23 Melting Point: 39-42° MDL No.: MFCD00048232

Formula: C₁₁H₁₄O₃
Melting Pt: 39…42 °C
Storage Temperature: Ambient
MDL Number: MFCD00048232
CAS Number: 122-48-5
EINECS: 204-548-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR