4-Pyridylacetonitrile hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Pyridin-4-yl)acetonitrile hydrochloride

B25095-03 B25095-06 B25095-14
CAAAAB25095-03EA 87.53 CAD
CAAAAB25095-03 CAAAAB25095-06 CAAAAB25095-14
4-Pyridylacetonitrile hydrochloride 98%
4-Pyridylacetonitrile hydrochloride

Purity: 98% CAS No.: 92333-25-0 Molecular Formula: C7H6N2.HCl Formula Weight: 154.60 Melting Point: ca 260° dec. UN No.: 3439 MDL No.: MFCD00012825

Formula: C₇H₇ClN₂
MW: 154.6 g/mol
Melting Pt: ∼ 260 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012825
CAS Number: 92333-25-0
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR