Methyl-3,5-dibromosalicylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3,5-dibromo-2-hydroxybenzoate

B25077-14 B25077-22
CAAAAB25077-14EA 121.19 CAD
CAAAAB25077-14 CAAAAB25077-22 CAAAAB25077-36
Methyl-3,5-dibromosalicylate 98%
Methyl 3,5-dibromosalicylate

Purity: 98% CAS No.: 21702-79-4 Molecular Formula: C8H6Br2O3 Formula Weight: 309.94 Melting Point: 150-152° MDL No.: MFCD00017534

Formula: C₈H₆Br₂O₃
MW: 309.94 g/mol
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00017534
CAS Number: 21702-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR