Ethyl-2-cyclohexylidenacetate 98%

Supplier: Thermo Scientific Chemicals

B25073-06 B25073-14 B25073-22
CAAAB25073-06EA 149.78 CAD
CAAAB25073-06 CAAAB25073-14 CAAAB25073-22
Ethyl-2-cyclohexylidenacetate 98%
Ethyl-2-cyclohexylidenacetate

Purity: 98% CAS No.: 1552-92-7 Molecular Formula: C10H16O2 Formula Weight: 168.24 Boiling Point: 48-49°/0.02mm MDL No.: MFCD00017627

Formula: C₁₀H₁₆O₂
Boiling Pt: 48…49 °C/0.02 mm
Storage Temperature: Ambient
MDL Number: MFCD00963857
CAS Number: 1552-92-7
EINECS: 216-297-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR