Formaldoxime trimer hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Danger

B25063-09 B25063-18
CAAAAB25063-09EA 657.28 CAD
CAAAAB25063-09 CAAAAB25063-18 CAAAB25063-22
Formaldoxime trimer hydrochloride 95%
Formaldoxime trimer hydrochloride

Purity: 95% CAS No.: 6286-29-9 Molecular Formula: C3H9N3O3.HCl Formula Weight: 171.57 Melting Point: ca 128° dec. Beilstein Reference: 3690722 UN No.: 2811 MDL No.: MFCD00012778

Formula: C₃H₉N₃O₃·HCl
Melting Pt: ∼ 128 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012778
CAS Number: 6286-29-9
EINECS: 222-440-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR