4-Azabenzimidazole 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 4H-Imidazo[4,5-b]pyridine

B25051-03 B25051-06 B25051-14
CAAAAB25051-03EA 114.55 CAD
CAAAAB25051-03 CAAAAB25051-06 CAAAAB25051-14
4-Azabenzimidazole 99%
4-Azabenzimidazole

Purity: 99% CAS No.: 273-21-2 Molecular Formula: C6H5N3 Formula Weight: 119.13 Melting Point: 145-150° Boiling Point: 140-150°/0.1mm Beilstein Reference: 2600 MDL No.: MFCD00005579

Formula: C₆H₅N₃
Boiling Pt: 140…150 °C/0.1 mm
Melting Pt: 145…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00005579
CAS Number: 273-21-2
EINECS: 205-987-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR