Imidazole-4-carboxaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

B25050-03 B25050-06 B25050-14
CAAAB25050-03EA 59.57 CAD
CAAAB25050-03 CAAAB25050-06 CAAAB25050-14
Imidazole-4-carboxaldehyde 97%
Imidazole-4-carboxaldehyde

Purity: 97% CAS No.: 3034-50-2 Molecular Formula: C4H4N2O Formula Weight: 96.09 Melting Point: 174-177°C MDL No.: MFCD00173726

Formula: C₄H₄N₂O
Melting Pt: 174…177°C
Storage Temperature: Ambient
MDL Number: MFCD00173726
CAS Number: 3034-50-2
EINECS: 221-227-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR