N-Boc-L-phenylalaninol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-(-)-BOC-Phenylalaninol, Boc-Phenylalaninol, (S)-tert-butyl 1-hydroxy-3-phenylpropan-2-ylcarbamate

B25019-03 B25019-06 B25019-14
CAAAAB25019-03EA 38.87 CAD
CAAAAB25019-03 CAAAAB25019-06 CAAAAB25019-14
N-Boc-L-phenylalaninol 99%
N-Boc-L-phenylalaninol

Purity: 99% CAS No.: 66605-57-0 Molecular Formula: C14H21NO3 Formula Weight: 251.33 Storage Temperature: 2°C to 8°C Melting Point: 95-98° Beilstein Reference: 3652004 Optical Rotation: -27.5° ± 2.5° (c=1 in methanol) MDL No.: MFCD00076976

Formula: C₁₄H₂₁NO₃
MW: 251.3 g/mol
Melting Pt: 95…98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00076976
CAS Number: 66605-57-0

Order Now

Specification Test Results

Optical rotation -27° (c=1 in methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR