3-Chloropropyl acetate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25013-14
B25013-22
B25013-36
CAAAAB25013-14EA
113.6
CAD
CAAAAB25013-14
CAAAAB25013-22
CAAAAB25013-36
3-Chloropropyl acetate 98%
3-Chloropropyl acetate
Purity: 98%
CAS No.: 628-09-1
Molecular Formula: CH3CO2(CH2)3Cl
Formula Weight: 136.58
Flash Point: 68°(154°F)
Refractive Index: 1.4290
Boiling Point: 70-72°/15mm
MDL No.: MFCD00001000
Formula:
C₅H₉ClO₂ Boiling Pt: 70…72 °C/15 mm Density: 1,12 Flash Pt: 68 °C(154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001000 CAS Number: 628-09-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...