1-Acetyl-4-fluoronaphthalene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4'-Fluoro-1'-acetonaphthone, 1-(4-Fluoronaphthalen-1-yl)ethanone, 1-(4-Fluoro-1-naphthyl)ethanone
B25004-03
B25004-14
CAAAAB25004-03EA
110.13
CAD
CAAAAB25004-03
CAAAAB25004-06
CAAAAB25004-14
1-Acetyl-4-fluoronaphthalene 97%
1-Acetyl-4-fluoronaphthalene
Purity: 97%
CAS No.: 316-68-7
Molecular Formula: C12H9FO
Formula Weight: 188.20
Flash Point:
Formula:
C₁₂H₉FO Boiling Pt: 164…166 °C/14 mm Density: 1,21 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00134475 CAS Number: 316-68-7 EINECS: 206-261-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...