5-Norbornene-2-carboxaldehyde (endo + exo mixture) 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Bicyclo[2.2.1]hept-5-ene-2-carboxaldehyde
B24973-14
B24973-22
CAAAAB24973-14EA
105.7
CAD
CAAAAB24973-14
CAAAAB24973-22
5-Norbornene-2-carboxaldehyde (endo + exo mixture) 95%
5-Norbornene-2-carboxaldehyde
Purity: 95%
CAS No.: 5453-80-5
Molecular Formula: C8H10O
Formula Weight: 122.17
Flash Point: 50°(122°F)
Refractive Index: 1.4910
Odor: Characteristic
Boiling Point: 68-70°/12mm
Beilstein Reference: 1363813
UN No.: 1989
MDL No.: MFCD00167568
Formula:
C₈H₁₀O MW: 122.17 g/mol Boiling Pt: 68…70 °C/12 mm Density: 1.032 Flash Pt: 50 °C (122 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00167568 CAS Number: 5453-80-5 EINECS: 226-698-9 UN: 1989 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...