Anthraquinone-2-sulfonic acid sodium salt, (H₂O 4-6%) 97% (dry weight)
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24969-0E
B24969-22
B24969-36
CAAAB24969-0EEA
720.48
CAD
CAAAB24969-0E
CAAAB24969-22
CAAAB24969-36
Anthraquinone-2-sulfonic acid sodium salt, (H₂O 4-6%) 97% (dry weight)
Anthraquinone-2-sulfonic acid sodium salt
Purity: 97%
CAS No.: 131-08-8
Molecular Formula: C14H7NaO5S.xH2O
Formula Weight: 310.26(anhy)
Melting Point:
Formula:
C₁₄H₇NaO₅S Melting Pt: >320 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001229 CAS Number: 131-08-8 EINECS: 205-009-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...