5-Iodo-2,4-dimethoxypyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning

B24967-01 B24967-03 B24967-06
CAAAAB24967-01EA 76.63 CAD
CAAAAB24967-01 CAAAAB24967-03 CAAAAB24967-06
5-Iodo-2,4-dimethoxypyrimidine 98%

Purity: 98% CAS No.: 52522-99-3 Molecular Formula: C6H7IN2O2 Formula Weight: 266.04 Melting Point: 71-75° MDL No.: MFCD00090865

Formula: C₆H₇IN₂O₂
Melting Pt: 71…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00090865
CAS Number: 52522-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR