Coniferyl alcohol 98%
Supplier: Thermo Scientific Chemicals
B24949-01
B24949-03
B24949-06
CAAAAB24949-01EA
199.08
CAD
CAAAAB24949-01
CAAAAB24949-03
CAAAAB24949-06
Coniferyl alcohol 98%
Coniferyl alcohol
Purity: 98%
CAS No.: 458-35-5
Molecular Formula: C10H12O3
Formula Weight: 180.20
Storage Temperature: -30°C to -10°C
Melting Point: 75-76°
Boiling Point: 163-165°/3mm
Beilstein Reference: 2048961
Merck Reference: 14,2504
MDL No.: MFCD00002922
Formula:
C₁₀H₁₂O₃ Boiling Pt: 163…165 °C/3 mm Melting Pt: 75…76 °C Storage Temperature: Freezer |
MDL Number:
MFCD00002922 CAS Number: 458-35-5 EINECS: 207-277-9 Merck Index: 14,2504 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...