2-(4-Hydroxyphenyl)propionic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B24938-09 B24938-18
CAAAAB24938-09EA 325.48 CAD
CAAAAB24938-09 CAAAAB24938-18
2-(4-Hydroxyphenyl)propionic acid 98%
2-(4-Hydroxyphenyl)propionic acid

Purity: 98% CAS No.: 938-96-5 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 130-132° MDL No.: MFCD00128847

Formula: C₉H₁₀O₃
Melting Pt: 130…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00128847
CAS Number: 938-96-5
EINECS: 213-352-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR