5-Bromo-2-iodobenzonitrile 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24880-06
B24880-14
CAAAAB24880-03EA
158.92
CAD
CAAAAB24880-03
CAAAAB24880-06
CAAAAB24880-14
5-Bromo-2-iodobenzonitrile 98+%
5-Bromo-2-iodobenzonitrile
Purity: 98+%
CAS No.: 121554-10-7
Molecular Formula: C7H3BrIN
Formula Weight: 307.92
Melting Point: 120-122°
Beilstein Reference: 4860282
UN No.: 3276
MDL No.: MFCD00672950
Formula:
C₇H₃BrIN Melting Pt: 120…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00672950 CAS Number: 121554-10-7 EINECS: 601-788-4 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...