Ethyl-4-phenylpyrrole-3-carboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24877-03
B24877-06
B24877-14
CAAAAB24877-03EA
130.67
CAD
CAAAAB24877-03
CAAAAB24877-06
CAAAAB24877-14
Ethyl-4-phenylpyrrole-3-carboxylate 97%
Ethyl-4-phenylpyrrole-3-carboxylate
Purity: 97%
CAS No.: 64276-62-6
Molecular Formula: C13H13NO2
Formula Weight: 215.25
Melting Point: 126-128°
MDL No.: MFCD03931521
Formula:
C₁₃H₁₃NO₂ Melting Pt: 126…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD03931521 CAS Number: 64276-62-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...