Ethyl-5-nitro-2-furoate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24861-03
B24861-06
CAAAAB24861-03EA
43.29
CAD
CAAAAB24861-03
CAAAAB24861-06
CAAAAB24861-14
Ethyl-5-nitro-2-furoate 97%
Ethyl-5-nitro-2-furoate
Purity: 97%
CAS No.: 943-37-3
Molecular Formula: C7H7NO5
Formula Weight: 185.14
Melting Point: 98-101°
MDL No.: MFCD00092566
Formula:
C₇H₇NO₅ Melting Pt: 98…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00092566 CAS Number: 943-37-3 EINECS: 213-400-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...