L-Glutamic acid 5-methyl ester 99%
Supplier: Thermo Scientific Chemicals
B24859-06
B24859-14
B24859-22
CAAAAB24859-06EA
54.19
CAD
CAAAAB24859-06
CAAAAB24859-14
CAAAAB24859-22
L-Glutamic acid 5-methyl ester 99%
L-Glutamic acid 5-methyl ester
Purity: 99%
CAS No.: 1499-55-4
Molecular Formula: C6H11NO4
Formula Weight: 161.16
Storage Temperature: 2°C to 8°C
Melting Point: 182° dec.
MDL No.: MFCD00002632
Formula:
C₆H₁₁NO₄ Melting Pt: 182 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002632 CAS Number: 1499-55-4 EINECS: 216-110-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...