6-Hydroxyhexanoic acid 95% may cont. variable amounts of dimer

Supplier: Thermo Scientific Chemicals
Warning

B24857-03 B24857-06 B24857-14
CAAAB24857-03EA 147.26 CAD
CAAAB24857-03 CAAAB24857-06 CAAAB24857-14
6-Hydroxyhexanoic acid 95% may cont. variable amounts of dimer
6-Hydroxyhexanoic acid

Purity: 95% CAS No.: 1191-25-9 Molecular Formula: C6H12O3 Formula Weight: 132.16 Storage Temperature: -30°C to -10°C Flash Point: 110°(230°F) Refractive Index: 1.4560 MDL No.: MFCD00046560

Formula: C₆H₁₂O₃
Melting Pt: 38…40 °C
Density: 1.08
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00046560
CAS Number: 1191-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR