Sodium phosphonoformate hexahydrate ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24784-03
B24784-06
B24784-14
CAAAAB24784-03EA
169.06
CAD
CAAAAB24784-03
CAAAAB24784-06
CAAAAB24784-14
Sodium phosphonoformate hexahydrate ≥98%
Sodium phosphonoformate hexahydrate
Purity: 98+%
CAS No.: 34156-56-4
Molecular Formula: Na2O3PCO2Na.6H2O
Formula Weight: 300.08 (191.98anhy)
MDL No.: MFCD00150176
Formula:
Na₂O₃PCO₂Na·6H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150176 CAS Number: 34156-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...