Tetrabutylammonium acetate 1.0 M in aqueous solution

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetra-n-butylammonium acetate

B24773-14 B24773-22
CAAAAB24773-14EA 78.84 CAD
CAAAAB24773-14 CAAAAB24773-22 CAAAAB24773-36
Tetrabutylammonium acetate 1.0 M in aqueous solution
Tetrabutylammonium acetate

Fieser: 3,277 9,442


CAS No.: 10534-59-5 Molecular Formula: C18H39NO2 Formula Weight: 301.51 Refractive Index: 1.3890 Boiling Point: 100° Beilstein Reference: 3599376

Formula: C₁₈H₃₉NO₂
MW: 301.51 g/mol
Boiling Pt: 100 °C
Density: 0.990
Storage Temperature: Ambient
MDL Number: MFCD00043208
CAS Number: 10534-59-5
EINECS: 234-101-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR