Ethyl hippurate 98%
Supplier: Thermo Scientific Chemicals
B24765-06
B24765-14
CAAAAB24765-06EA
113.76
CAD
CAAAAB24765-06
CAAAAB24765-14
CAAAAB24765-22
Ethyl hippurate 98%
Ethyl hippurate
Purity: 98%
CAS No.: 1499-53-2
Molecular Formula: C6H5CONHCH2CO2CH2CH3
Formula Weight: 207.23
Melting Point: 58-60°
MDL No.: MFCD00026890
Formula:
C₁₁H₁₃NO₃ Melting Pt: 58…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00026890 CAS Number: 1499-53-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...