3-Ethoxy-4-methoxybenzaldehyde 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Apremilast Impurity 28
B24762-06
B24762-14
CAAAAB24762-06EA
77.26
CAD
CAAAAB24762-06
CAAAAB24762-14
CAAAAB24762-22
3-Ethoxy-4-methoxybenzaldehyde 99%
3-Ethoxy-4-methoxybenzaldehyde
Purity: 99%
CAS No.: 1131-52-8
Molecular Formula: C10H12O3
Formula Weight: 180.20
Melting Point: 51-53°
MDL No.: MFCD00010128
Formula:
C₁₀H₁₂O₃ Melting Pt: 51…53 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010128 CAS Number: 1131-52-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...