2,2'-Thenil 98%

Supplier: Thermo Scientific Chemicals
Warning

B24753-03 B24753-06
CAAAAB24753-03EA 75.84 CAD
CAAAAB24753-03 CAAAAB24753-06
2,2'-Thenil 98%
2,2'-Thenil

Purity: 98% CAS No.: 7333-07-5 Molecular Formula: C10H6O2S2 Formula Weight: 222.29 Melting Point: 81-84° MDL No.: MFCD00173678

Formula: C₁₀H₆O₂S₂
Melting Pt: 81…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00173678
CAS Number: 7333-07-5
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR