Methyl-1,2,4-triazole-3-carboxylate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 1H-1,2,4-triazole-3-carboxylate, Methyl 1H-1,2,4-triazole-5-carboxylate
B24750-06
B24750-14
B24750-22
CAAAAB24750-06EA
66.83
CAD
CAAAAB24750-06
CAAAAB24750-14
CAAAAB24750-22
Methyl-1,2,4-triazole-3-carboxylate 98%
Methyl 1,2,4-triazole-3-carboxylate
Purity: 98%
CAS No.: 4928-88-5
Molecular Formula: C4H5N3O2
Formula Weight: 127.10
MDL No.: MFCD00135989
Formula:
C₄H₅N₃O₂ MW: 127.10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00135989 CAS Number: 4928-88-5 EINECS: 480-470-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...