DL-Penicillamine 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24710-03
B24710-06
CAAAAB24710-03EA
53.4
CAD
CAAAAB24710-03
CAAAAB24710-06
CAAAAB24710-14
DL-Penicillamine 97+%
DL-Penicillamine
Heavy metal chelating agent.
Purity: 97+%
CAS No.: 52-66-4
Molecular Formula: C5H11NO2S
Formula Weight: 149.21
Storage Temperature: 2°C to 8°C
Odor: Unpleasant
Beilstein Reference: 2039817
Merck Reference: 14,7088
MDL No.: MFCD00004856
Formula:
C₅H₁₁NO₂S Melting Pt: ca 212 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004856 CAS Number: 52-66-4 EINECS: 200-147-2 Merck Index: 14,7088 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...