Sodium hydrogen fumarate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Monosodium Fumarate, Fumaric Acid Monosodium Salt

B24683-18 B24683-30
CAAAAB24683-18EA 65.57 CAD
CAAAAB24683-18 CAAAAB24683-30
Sodium hydrogen fumarate 98%
Sodium hydrogen fumarate

Purity: 98% CAS No.: 5873-57-4 Molecular Formula: C4H3NaO4 Formula Weight: 138.06 MDL No.: MFCD00039097

Formula: C₄H₄O₄
MW: 138.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00039097
CAS Number: 5873-57-4
EINECS: 227-535-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR