Benzyl-3-hydroxyphenylacetate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Benzyl 2-(3-hydroxyphenyl)acetate

B24679-06 B24679-14
CAAAAB24679-03EA 32.45 CAD
CAAAAB24679-03 CAAAAB24679-06 CAAAAB24679-14
Benzyl-3-hydroxyphenylacetate 98%
Benzyl-3-hydroxyphenylacetate

Purity: 98% CAS No.: 295320-25-1 Molecular Formula: C15H14O3 Formula Weight: 242.27 Melting Point: 79-81° MDL No.: MFCD04039768

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Melting Pt: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD04039768
CAS Number: 295320-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR