1-(2-Aminoethyl)pyrrolidine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(2-Aminoethyl)pyrrolidine, Pyrrolidinoethylamine

B24645-06 B24645-14 B24645-22
CAAAB24645-06EA 90.69 CAD
CAAAB24645-06 CAAAB24645-14 CAAAB24645-22
1-(2-Aminoethyl)pyrrolidine 99%
1-(2-Aminoethyl)pyrrolidine

Purity: 99% CAS No.: 7154-73-6 Molecular Formula: C6H14N2 Formula Weight: 114.19 Flash Point: 47°(116°F) Refractive Index: 1.4687 Odor: Amine-like Boiling Point: 161° Beilstein Reference: 102978 UN No.: 2734 MDL No.: MFCD00003182

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 161 °C
Density: 0.904
Flash Pt: 47 °C(116 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003182
CAS Number: 7154-73-6
EINECS: 230-509-5
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR