Suberoyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

B24580-06 B24580-14
CAAAB24580-06EA 124.82 CAD
CAAAB24580-06 CAAAB24580-14
Suberoyl chloride 97%
Suberoyl chloride

Purity: 97% CAS No.: 10027-07-3 Molecular Formula: C8H12Cl2O2 Formula Weight: 211.09 Flash Point: 110°(230°F) Refractive Index: 1.4680 Boiling Point: 162-163°/15mm UN No.: 3265 MDL No.: MFCD00000764

Formula: C₈H₁₂Cl₂O₂
Boiling Pt: 162…163 °C/15 mm
Density: 1.172
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000764
CAS Number: 10027-07-3
UN: 1760
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR