Iron(II) gluconate hydrate 94%
Supplier: Thermo Scientific Chemicals
B24561-22
B24561-36
CAAAAB24561-22EA
36.02
CAD
CAAAAB24561-22
CAAAAB24561-36
Iron(II) gluconate hydrate 94%
Iron(II) gluconate hydrate
Purity: 94%
CAS No.: 22830-45-1
Molecular Formula: C12H22FeO14.2H2O
Formula Weight: 482.19 (446.19anhy)
Melting Point: ca 140°
MDL No.: MFCD00150872
Formula:
C₁₂H₂₄FeO₁₅ Melting Pt: ∼ 140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150872 CAS Number: 22830-45-1 EINECS: 206-076-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...