D-Asparagine monohydrate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
(R)-2-Aminosuccinamic acid hydrate, D-2-Aminosuccinamic acid, H-D-Asn-OH·H₂O, (R)-2,4-Diamino-4-oxobutanoic acid hydrate
B24556-14
B24556-22
CAAAB24556-14EA
61.15
CAD
CAAAB24556-14
CAAAB24556-22
D-Asparagine monohydrate 99%
D-Asparagine monohydrate
Purity: 99%
CAS No.: 5794-24-1
Molecular Formula: C4H8N2O3.H2O
Formula Weight: 150.14 (132.12anhy)
Melting Point: 274-276° dec.
MDL No.: MFCD00149558
Formula:
C₄H₁₀N₂O₄ MW: 150.14 g/mol (132.12 anhy) Melting Pt: 274…276 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149558 CAS Number: 5794-24-1 Merck Index: 14,837 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...