4-Hydroxybenzoylacrylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24519-06
B24519-14
CAAAB24519-03EA
182.62
CAD
CAAAB24519-03
CAAAB24519-06
CAAAB24519-14
4-Hydroxybenzoylacrylic acid 97%
4-Hydroxybenzoylacrylic acid
Purity: 97%
CAS No.: 24849-48-7
Molecular Formula: C10H8O4
Formula Weight: 192.17
Melting Point: 197-198°
MDL No.: MFCD00460607
Formula:
C₁₀H₈O₄ Melting Pt: 197…198 °C Storage Temperature: Ambient |
MDL Number:
MFCD00460607 CAS Number: 24849-48-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...