3-Fluoro-4-methylphenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24512-03
B24512-06
B24512-14
CAAAB24512-03EA
46.93
CAD
CAAAB24512-03
CAAAB24512-06
CAAAB24512-14
3-Fluoro-4-methylphenylboronic acid 98%
3-Fluoro-4-methylphenylboronic acid
Purity: 98%
CAS No.: 168267-99-0
Molecular Formula: C7H8BFO2
Formula Weight: 153.95
Melting Point: 234°
Beilstein Reference: 8544594
Formula:
C₇H₈BFO₂ Melting Pt: 234 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683115 CAS Number: 168267-99-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...