Choline hydroxide 46% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Hydroxyethyltrimethylammonium Hydroxide, Choline

B24443-AE B24443-AP
CAAAB24443-AEEA 136.99 CAD
CAAAB24443-AE CAAAB24443-AP
Choline hydroxide 46% (w/w) in aqueous solution
Choline hydroxide

CAS No.: 123-41-1 Molecular Formula: C5H15NO2 Formula Weight: 121.18 Refractive Index: 1.4270 UN No.: 3267 MDL No.: MFCD00002831

Formula: C₅H₁₅NO₂
MW: 121.18 g/mol
Density: 1.073
Storage Temperature: Ambient
MDL Number: MFCD00002831
CAS Number: 123-41-1
EINECS: 204-625-1
UN: 3267
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR