4-tert-Butylphenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-tert-Butylbenzeneboronic acid, (4-(tert-Butyl)phenyl)boronic acid
B24408-03
B24408-06
CAAAB24408-03EA
44.24
CAD
CAAAB24408-03
CAAAB24408-06
4-tert-Butylphenylboronic acid 97%
4-tert-Butylphenylboronic acid
Purity: 97%
CAS No.: 123324-71-0
Molecular Formula: C10H15BO2
Formula Weight: 178.04
Storage Temperature: 2°C to 8°C
Melting Point: 191-196°
Beilstein Reference: 5330959
MDL No.: MFCD01009697
Formula:
C₁₀H₁₅BO₂ MW: 178.04 g/mol Melting Pt: 191…196 °C Storage Temperature: Ambient |
MDL Number:
MFCD01009697 CAS Number: 123324-71-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...